2,4-Difluoro-5-nitrobenzyl alcohol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2,4-Difluoro-5-nitrophenyl)methanol
PC51225-1G
PC51225-5G
APOSPC51225-1GEA
36.4
EUR
APOSPC51225-1G
APOSPC51225-5G
2,4-Difluoro-5-nitrobenzyl alcohol 97%
2,4-Difluoro-5-nitrobenzyl alcohol
Formel:
C₇H₅F₂NO₃ Molecular Weight: 189,116 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12047726 CAS-Nummer: 935287-88-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...