2-Hydroxyethyl trifluoromethyl ether 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Trifluoromethoxy)ethanol, 2-(Trifluoromethoxy)ethan-1-ol

PC5131-1G PC5131-250MG PC5131-500MG
APOSPC5131-1GEA 683 EUR
APOSPC5131-1G APOSPC5131-250MG APOSPC5131-500MG
2-Hydroxyethyl trifluoromethyl ether 95%
2-Hydroxyethyl trifluoromethyl ether
Formel: C₃H₅F₃O₂
Molecular Weight: 130,066 g/mol
Siedepunkt: 91 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11858187
CAS-Nummer: 362631-84-3
UN: 3271
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR