6-Fluoro-5-iodopyridin-2-ol 95%

Lieferant: Apollo Scientific
Achtung

PC51320-1G PC51320-250MG PC51320-5G
APOSPC51320-1GEA 296 EUR
APOSPC51320-1G APOSPC51320-250MG APOSPC51320-5G
6-Fluoro-5-iodopyridin-2-ol 95%
6-Fluoro-5-iodopyridin-2-ol
Formel: C₅H₃FINO
Molecular Weight: 238,986 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16611044
CAS-Nummer: 884660-48-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR