6-Fluoro-5-iodopyridin-2-amine 95%

Lieferant: Apollo Scientific
Achtung

PC51322-1G PC51322-250MG PC51322-5G
APOSPC51322-1GEA 72.5 EUR
APOSPC51322-1G APOSPC51322-250MG APOSPC51322-5G
6-Fluoro-5-iodopyridin-2-amine 95%
6-Fluoro-5-iodopyridin-2-amine
Formel: C₅H₄FIN₂
Molecular Weight: 238,002 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09032812
CAS-Nummer: 884660-47-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR