4-(Heptafluoroisopropyl)iodobenzene 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(1,1,1,2,3,3,3-Heptafluoropropan-2-yl)-4-iodobenzene

PC51344-1G PC51344-250MG PC51344-5G
APOSPC51344-1GEA 632 EUR
APOSPC51344-1G APOSPC51344-250MG APOSPC51344-5G
4-(Heptafluoroisopropyl)iodobenzene 95%
4-(Heptafluoroisopropyl)iodobenzene
Formel: C₉H₄F₇I
Molecular Weight: 372,021 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25372105
CAS-Nummer: 40161-59-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR