2-Bromo-3,6-difluorobenzotrifluoride 97%
Lieferant: Apollo Scientific
Synonyme:
2-Bromo-1,4-difluoro-3-(trifluoromethyl)benzene
PC51364-1G
PC51364-5G
PC51364-250MG
APOSPC51364-1GEA
412
EUR
APOSPC51364-1G
APOSPC51364-5G
APOSPC51364-250MG
2-Bromo-3,6-difluorobenzotrifluoride 97%
2-Bromo-3,6-difluorobenzotrifluoride
Formel:
C₇H₂BrF₅ Molecular Weight: 260,987 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16817690 CAS-Nummer: 1242339-93-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...