2-(3,4-Difluoro-2-methoxyphenyl)acetic acid 95%

Lieferant: Apollo Scientific

PC51390-1G PC51390-250MG
APOSPC51390-1GEA 609 EUR
APOSPC51390-1G APOSPC51390-250MG
2-(3,4-Difluoro-2-methoxyphenyl)acetic acid 95%
2-(3,4-Difluoro-2-methoxyphenyl)acetic acid
Formel: C₉H₈F₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28626908
CAS-Nummer: 1558274-26-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR