3,4-Difluoro-2-methoxybenzyl bromide 95%

Lieferant: Apollo Scientific

PC51393-1G PC51393-250MG PC51393-5G
APOSPC51393-1GEA 463 EUR
APOSPC51393-1G APOSPC51393-250MG APOSPC51393-5G
3,4-Difluoro-2-methoxybenzyl bromide 95%
3,4-Difluoro-2-methoxybenzyl bromide
Formel: C₈H₇BrF₂O
Molecular Weight: 237.041 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28789284
CAS-Nummer: 1804416-43-0
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR