4,5-Diamino-2-fluorobenzotrifluoride 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,2-Diamino-4-fluoro-5-(trifluoromethyl)benzene, 4-Fluoro-5-(trifluoromethyl)benzene-1,2-diamine

PC5171-1G PC5171-5G PC5171-10G
APOSPC5171-1GEA 140 EUR
APOSPC5171-1G APOSPC5171-5G APOSPC5171-10G
4,5-Diamino-2-fluorobenzotrifluoride 97%
4,5-Diamino-2-fluorobenzotrifluoride
Formel: C₇H₆F₄N₂
Molecular Weight: 194,13 g/mol
Schmelzpunkt: 48…51 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04038250
CAS-Nummer: 179062-06-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR