3-Chloro-4-iodobenzyl fluoride

Lieferant: Apollo Scientific

PC51814-1G PC51814-250MG
APOSPC51814-1GEA 574 EUR
APOSPC51814-1G APOSPC51814-250MG
3-Chloro-4-iodobenzyl fluoride
3-Chloro-4-iodobenzyl fluoride
Formel: C₇H₅ClFI
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2414146-35-1
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR