2-Chloro-5-fluoro-4-nitroaniline 97%

Lieferant: Apollo Scientific
Achtung

PC5182-1G PC5182-10G PC5182-5G
APOSPC5182-1GEA 145 EUR
APOSPC5182-1G APOSPC5182-10G APOSPC5182-5G
2-Chloro-5-fluoro-4-nitroaniline 97%
2-Chloro-5-fluoro-4-nitroaniline
Formel: C₆H₄ClFN₂O₂
Molecular Weight: 190.56 g/mol
Schmelzpunkt: 128…131 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094272
CAS-Nummer: 517920-71-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR