4-Hydroxy-3-nitrophenylsulphur pentafluoride 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Nitro-4-(pentafluorothio)phenol, 2-Hydroxy-5-(pentafluorothio)nitrobenzene

PC5199-1G PC5199-250MG PC5199-5G
APOSPC5199-1GEA 183 EUR
APOSPC5199-1G APOSPC5199-250MG APOSPC5199-5G
4-Hydroxy-3-nitrophenylsulphur pentafluoride 95%
4-Hydroxy-3-nitrophenylsulphur pentafluoride
Formel: C₆H₄F₅NO₃S
Molecular Weight: 265,158 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11501052
CAS-Nummer: 1159512-26-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR