Ethyl 5-amino-4-bromo-1-(2,2,2-trifluoroethyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

PC520002-1G
APOSPC520002-1GEA 912 EUR
APOSPC520002-1G
Ethyl 5-amino-4-bromo-1-(2,2,2-trifluoroethyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-bromo-1-(2,2,2-trifluoroethyl)pyrazole-3-carboxylate
Formel: C₈H₉BrF₃N₃O₂
Molecular Weight: 316.075 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23135207
CAS-Nummer: 1427023-16-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR