Tris(2,2,3,3-tetrafluoropropyl) borate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Tris(2,2,3,3-tetrafluoropropoxy)borane
PC52051-5G
APOSPC52051-5GEA
607
EUR
APOSPC52051-5G
Tris(2,2,3,3-tetrafluoropropyl) borate
Tris(2,2,3,3-tetrafluoropropyl) borate
Formel:
C₉H₉BF₁₂O₃ Molecular Weight: 403,958 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22124055 CAS-Nummer: 2317-76-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...