4,4,4-Trifluoro-1-(4-pyridyl)butane-1,3-dione 95%
Lieferant: Apollo Scientific
PC520516-1G
PC520516-5G
APOSPC520516-1GEA
118
EUR
APOSPC520516-1G
APOSPC520516-5G
4,4,4-Trifluoro-1-(4-pyridyl)butane-1,3-dione 95%
4,4,4-Trifluoro-1-(4-pyridyl)butane-1,3-dione
Formel:
C₉H₆F₃NO₂ Molecular Weight: 217,145 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01566518 CAS-Nummer: 399-06-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...