5-(4-Fluoro-benzyl)-thiazol-2-ylamine 97%

Lieferant: Apollo Scientific

PC520520-1G PC520520-5G
APOSPC520520-1GEA 280 EUR
APOSPC520520-1G APOSPC520520-5G
5-(4-Fluoro-benzyl)-thiazol-2-ylamine 97%
5-(4-Fluoro-benzyl)-thiazol-2-ylamine
Formel: C₁₀H₉FN₂S
Molecular Weight: 208,255 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02214714
CAS-Nummer: 315241-39-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR