4-Chloro-3-(trifluoromethyl)benzyl chloride 98%

Lieferant: Apollo Scientific

Synonyme: 1-Chloro-4-(chloromethyl)-2-(trifluoromethyl)benzene

PC520528-1G PC520528-25G PC520528-5G
APOSPC520528-1GEA 52.2 EUR
APOSPC520528-1G APOSPC520528-25G APOSPC520528-5G
4-Chloro-3-(trifluoromethyl)benzyl chloride 98%
4-Chloro-3-(trifluoromethyl)benzyl chloride
Formel: C₈H₅Cl₂F₃
Molecular Weight: 229,027 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03412205
CAS-Nummer: 23131-73-9
UN: 1760
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR