5-Amino-2,3-difluorobenzeneboronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Borono-4,5-difluoroaniline

PC52053-1G PC52053-250MG
APOSPC52053-1GEA 764 EUR
APOSPC52053-1G APOSPC52053-250MG
5-Amino-2,3-difluorobenzeneboronic acid
5-Amino-2,3-difluorobenzeneboronic acid
Formel: C₆H₆BF₂NO₂
Molecular Weight: 172,925 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11855852
CAS-Nummer: 1150114-58-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR