3-Bromo-2,4,5,6-tetrafluorobenzotrifluoride
Lieferant: Apollo Scientific
PC520571-10G
PC520571-1G
PC520571-5G
APOSPC520571-10GEA
1550
EUR
APOSPC520571-10G
APOSPC520571-1G
APOSPC520571-5G
3-Bromo-2,4,5,6-tetrafluorobenzotrifluoride
3-Bromo-2,4,5,6-tetrafluorobenzotrifluoride
Formel:
C₇BrF₇ Molecular Weight: 296,968 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09037800 CAS-Nummer: 113601-46-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...