Dimethyl 2-(3-trifluoromethylbenzyl)malonate 97%

Lieferant: Apollo Scientific

PC520575-1G PC520575-5G
APOSPC520575-1GEA 199 EUR
APOSPC520575-1G APOSPC520575-5G
Dimethyl 2-(3-trifluoromethylbenzyl)malonate 97%
Dimethyl 2-(3-trifluoromethylbenzyl)malonate
Formel: C₁₃H₁₃F₃O₄
Molecular Weight: 290.235 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07780246
CAS-Nummer: 1186194-84-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR