Ethyl-4-fluoroindole-2-carboxylate 97%

Lieferant: Apollo Scientific

Synonyme: Ethyl 4-fluoro-1H-indole-2-carboxylate

PC520581-1G PC520581-500MG
APOSPC520581-1GEA 686 EUR
APOSPC520581-1G APOSPC520581-500MG
Ethyl-4-fluoroindole-2-carboxylate 97%
Ethyl-4-fluoroindole-2-carboxylate
Formel: C₁₁H₁₀FNO₂
Molecular Weight: 207,201 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09264533
CAS-Nummer: 348-32-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR