5-(Trifluoromethoxy)pyridine-2-carboxylic acid 95%

Lieferant: Apollo Scientific

Synonyme: 5-(Trifluoromethoxy)picolinic acid

PC520601-1G PC520601-250MG
APOSPC520601-1GEA 1050 EUR
APOSPC520601-1G APOSPC520601-250MG
5-(Trifluoromethoxy)pyridine-2-carboxylic acid 95%
5-(Trifluoromethoxy)pyridine-2-carboxylic acid
Formel: C₇H₄F₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD10697540
CAS-Nummer: 102771-66-4
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR