Methyl 5-bromo-2-(difluoromethoxy)benzoate 95%
Lieferant: Apollo Scientific
PC520627-1G
PC520627-5G
APOSPC520627-1GEA
119
EUR
APOSPC520627-1G
APOSPC520627-5G
Methyl 5-bromo-2-(difluoromethoxy)benzoate 95%
Methyl 5-bromo-2-(difluoromethoxy)benzoate
Formel:
C₉H₇BrF₂O₃ Molecular Weight: 281,051 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11110847 CAS-Nummer: 1131587-78-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...