2-Amino-4-fluorobenzaldehyde 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Fluoroanthranilaldehyde, 5-Fluoro-2-formylaniline

PC52065-1G PC52065-250MG PC52065-5G
APOSPC52065-1GEA 71.2 EUR
APOSPC52065-1G APOSPC52065-250MG APOSPC52065-5G
2-Amino-4-fluorobenzaldehyde 98%
2-Amino-4-fluorobenzaldehyde
Formel: C₇H₆FNO
Molecular Weight: 139.127 g/mol
Schmelzpunkt: 49…52 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD10000632
CAS-Nummer: 152367-89-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR