3-(Trifluoromethyl)pyridazine 97%

Lieferant: Apollo Scientific

PC520717-10G PC520717-1G PC520717-5G
APOSPC520717-10GEA 1520 EUR
APOSPC520717-10G APOSPC520717-1G APOSPC520717-5G
3-(Trifluoromethyl)pyridazine 97%
3-(Trifluoromethyl)pyridazine
Formel: C₅H₃F₃N₂
Molecular Weight: 148.086 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16140190
CAS-Nummer: 132537-43-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR