3-Bromo-2-hydroxy-4-(trifluoromethyl)pyridine 95%

Lieferant: Apollo Scientific

PC520727-250MG PC520727-500MG
APOSPC520727-250MGEA 280 EUR
APOSPC520727-250MG APOSPC520727-500MG
3-Bromo-2-hydroxy-4-(trifluoromethyl)pyridine 95%
3-Bromo-2-hydroxy-4-(trifluoromethyl)pyridine
Formel: C₆H₃BrF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD16610037
CAS-Nummer: 1227591-86-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR