2-Amino-3-fluoro-4-(trifluoromethyl)pyridine 97%

Lieferant: Apollo Scientific

Synonyme: 3-Fluoro-4-(trifluoromethyl)pyridin-2-amine

PC520730-1G PC520730-500MG PC520730-5G
APOSPC520730-1GEA 61.3 EUR
APOSPC520730-1G APOSPC520730-500MG APOSPC520730-5G
2-Amino-3-fluoro-4-(trifluoromethyl)pyridine 97%
2-Amino-3-fluoro-4-(trifluoromethyl)pyridine
Formel: C₆H₄F₄N₂
Molecular Weight: 180.103 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16606170
CAS-Nummer: 1227514-27-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR