1-(Bromomethyl)-4-(1,1,2,2-tetrafluoroethoxy)benzene 95%

Lieferant: Apollo Scientific

PC520733-25G PC520733-50G
APOSPC520733-25GEA 605 EUR
APOSPC520733-25G APOSPC520733-50G
1-(Bromomethyl)-4-(1,1,2,2-tetrafluoroethoxy)benzene 95%
1-(Bromomethyl)-4-(1,1,2,2-tetrafluoroethoxy)benzene
Formel: C₉H₇BrF₄O
Molecular Weight: 287,049 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16619616
CAS-Nummer: 67033-41-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR