1-(Bromomethyl)-1-(trifluoromethyl)cyclopropane 95%

Lieferant: Apollo Scientific
Achtung

PC520750-1G PC520750-250MG
APOSPC520750-1GEA 696 EUR
APOSPC520750-1G APOSPC520750-250MG
1-(Bromomethyl)-1-(trifluoromethyl)cyclopropane 95%
1-(Bromomethyl)-1-(trifluoromethyl)cyclopropane
Formel: C₅H₆BrF₃
Molecular Weight: 203 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD20230586
CAS-Nummer: 1155272-93-7
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR