2-Chloro-6-iodo-4-(trifluoromethyl)aniline 97%
Lieferant: Apollo Scientific
PC520752-10G
PC520752-1G
PC520752-5G
APOSPC520752-10GEA
1040
EUR
APOSPC520752-10G
APOSPC520752-1G
APOSPC520752-5G
2-Chloro-6-iodo-4-(trifluoromethyl)aniline 97%
2-Chloro-6-iodo-4-(trifluoromethyl)aniline
Formel:
C₇H₄ClF₃IN Molecular Weight: 321,466 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20492612 CAS-Nummer: 1065102-88-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...