Benzene-1,3-disulphonyl fluoride 95%

Lieferant: Apollo Scientific

PC520783-1G PC520783-5G
APOSPC520783-1GEA 187 EUR
APOSPC520783-1G APOSPC520783-5G
Benzene-1,3-disulphonyl fluoride 95%
Benzene-1,3-disulphonyl fluoride
Formel: C₆H₄F₂O₄S₂
Molecular Weight: 242,22 g/mol
Schmelzpunkt: 37…42 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD28346978
CAS-Nummer: 7552-55-8
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR