3-Fluoropiperidine-1-sulphonyl chloride 95%

Lieferant: Apollo Scientific

PC520788-1G PC520788-250MG PC520788-500MG
APOSPC520788-1GEA 582 EUR
APOSPC520788-1G APOSPC520788-250MG APOSPC520788-500MG
3-Fluoropiperidine-1-sulphonyl chloride 95%
3-Fluoropiperidine-1-sulphonyl chloride
Formel: C₅H₉ClFNO₂S
Molecular Weight: 201.647 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29925539
CAS-Nummer: 1845693-87-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR