3,3-Difluoropyrrolidine-1-sulphonyl chloride 95%

Lieferant: Apollo Scientific

PC520803-250MG PC520803-500MG
APOSPC520803-250MGEA 861 EUR
APOSPC520803-250MG APOSPC520803-500MG
3,3-Difluoropyrrolidine-1-sulphonyl chloride 95%
3,3-Difluoropyrrolidine-1-sulphonyl chloride
Formel: C₄H₆ClF₂NO₂S
Molecular Weight: 205,611 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27918725
CAS-Nummer: 1187160-31-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR