3-Phenyl-5-(trifluoromethyl)-1H-1,2,4-triazole 95%

Lieferant: Apollo Scientific

PC520806-1G PC520806-250MG PC520806-500MG
APOSPC520806-1GEA 640 EUR
APOSPC520806-1G APOSPC520806-250MG APOSPC520806-500MG
3-Phenyl-5-(trifluoromethyl)-1H-1,2,4-triazole 95%
3-Phenyl-5-(trifluoromethyl)-1H-1,2,4-triazole
Formel: C₉H₆F₃N₃
Molecular Weight: 213,159 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07779829
CAS-Nummer: 60406-64-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR