1-(Trifluoromethyl)cyclopropanamine hydrochloride 95%
Lieferant: Apollo Scientific
PC520815-250MG
PC520815-500MG
APOSPC520815-250MGEA
120
EUR
APOSPC520815-250MG
APOSPC520815-500MG
1-(Trifluoromethyl)cyclopropanamine hydrochloride 95%
1-(Trifluoromethyl)cyclopropanamine hydrochloride
Formel:
C₄H₆F₃N·ClH Molecular Weight: 161,553 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11109575 CAS-Nummer: 112738-67-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...