3,5-Difluoro-4-(trifluoromethyl)pyridine 95%
Lieferant: Apollo Scientific
PC520823-1G
PC520823-500MG
PC520823-5G
APOSPC520823-1GEA
338
EUR
APOSPC520823-1G
APOSPC520823-500MG
APOSPC520823-5G
3,5-Difluoro-4-(trifluoromethyl)pyridine 95%
3,5-Difluoro-4-(trifluoromethyl)pyridine
Formel:
C₆H₂F₅N Lagertemperatur: Raumtemperatur |
MDL:
MFCD22378815 CAS-Nummer: 1391033-20-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...