(3R)-3-(Trifluoromethyl)piperidine 97%

Lieferant: Apollo Scientific

PC520826-1G PC520826-500MG PC520826-100MG PC520826-250MG
APOSPC520826-1GEA 516 EUR
APOSPC520826-1G APOSPC520826-500MG APOSPC520826-100MG APOSPC520826-250MG
(3R)-3-(Trifluoromethyl)piperidine 97%
(3R)-3-(Trifluoromethyl)piperidine
Formel: C₆H₁₀F₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD22989486
CAS-Nummer: 1282530-77-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR