4-Fluoro-3-methylphenyl isocyanate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Fluoro-4-isocyanato-2-methylbenzene

PC52086-1G PC52086-5G PC52086-25G
APOSPC52086-1GEA 55.1 EUR
APOSPC52086-1G APOSPC52086-5G APOSPC52086-25G
4-Fluoro-3-methylphenyl isocyanate 98%
4-Fluoro-3-methylphenyl isocyanate

May form small amount of sediment

Formel: C₈H₆FNO
Molecular Weight: 151,138 g/mol
Siedepunkt: 186 °C
Dichte: 1,172 g/cm³ (25 °C)
Flammpunkt: 57 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03427210
CAS-Nummer: 351003-65-1
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR