2-Chloro-3,5-bis(trifluoromethyl)pyridine 95%

Lieferant: Apollo Scientific
Gefahr

PC520891-1G PC520891-5G
APOSPC520891-1GEA 678 EUR
APOSPC520891-1G APOSPC520891-5G
2-Chloro-3,5-bis(trifluoromethyl)pyridine 95%
2-Chloro-3,5-bis(trifluoromethyl)pyridine
Formel: C₇H₂ClF₆N
Molecular Weight: 249.541 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00082555
CAS-Nummer: 70158-60-0
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR