2-Cyano-6-(trifluoromethyl)pyridine 97%
Lieferant: Apollo Scientific
Synonyme:
6-(Trifluoromethyl)picolinonitrile
PC520913-5G
APOSPC520913-5GEA
122
EUR
APOSPC520913-5G
2-Cyano-6-(trifluoromethyl)pyridine 97%
2-Cyano-6-(trifluoromethyl)pyridine
Formel:
C₇H₃F₃N₂ Molecular Weight: 172,107 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368048 CAS-Nummer: 887583-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...