2-Bromo-5-fluoro-3-nitrobenzyl alcohol

Lieferant: Apollo Scientific
Achtung

PC52102-1G PC52102-5G PC52102-250MG
APOSPC52102-1GEA 261 EUR
APOSPC52102-1G APOSPC52102-5G APOSPC52102-250MG
2-Bromo-5-fluoro-3-nitrobenzyl alcohol
2-Bromo-5-fluoro-3-nitrobenzyl alcohol
Formel: C₇H₅BrFNO₃
Molecular Weight: 250,022 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28740116
CAS-Nummer: 1805109-31-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR