4-Fluoro-3-iodo-2-methoxypyridine 95%

Lieferant: Apollo Scientific
Achtung

PC52155-1G PC52155-250MG PC52155-5G
APOSPC52155-1GEA 632 EUR
APOSPC52155-1G APOSPC52155-250MG APOSPC52155-5G
4-Fluoro-3-iodo-2-methoxypyridine 95%
4-Fluoro-3-iodo-2-methoxypyridine
Formel: C₆H₅FINO
Molecular Weight: 253,013 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28796405
CAS-Nummer: 1806345-85-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR