3-Bromo-4-(trifluoromethoxy)benzal fluoride
Lieferant: Apollo Scientific
|
Achtung
|
PC52180-1G
PC52180-250MG
PC52180-5G
APOSPC52180-1GEA
0
EUR
APOSPC52180-1G
APOSPC52180-250MG
APOSPC52180-5G
3-Bromo-4-(trifluoromethoxy)benzal fluoride
3-Bromo-4-(trifluoromethoxy)benzal fluoride
Formel:
C₈H₄BrF₅O Lagertemperatur: Raumtemperatur |
MDL:
MFCD31812117 CAS-Nummer: 2166881-48-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...