4-Bromo-2,3-difluorobenzal fluoride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-4-(difluoromethyl)-2,3-difluorobenzene
PC52186-1G
PC52186-250MG
APOSPC52186-1GEA
522
EUR
APOSPC52186-1G
APOSPC52186-250MG
4-Bromo-2,3-difluorobenzal fluoride
4-Bromo-2,3-difluorobenzal fluoride
Formel:
C₇H₃BrF₄ Molecular Weight: 242,996 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28737889 CAS-Nummer: 1449290-34-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...