1,1,1,3,3,3-Hexafluoro-2-(3-iodophenyl)propan-2-ol 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC52192-1G
PC52192-250MG
APOSPC52192-1GEA
174
EUR
APOSPC52192-1G
APOSPC52192-250MG
1,1,1,3,3,3-Hexafluoro-2-(3-iodophenyl)propan-2-ol 95%
1,1,1,3,3,3-Hexafluoro-2-(3-iodophenyl)propan-2-ol
Formel:
C₉H₅F₆IO Molecular Weight: 370,03 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31922516 CAS-Nummer: 2366994-34-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...