5-(Difluoromethyl)pyridine-2-carbaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-(Difluoromethyl)picolinaldehyde
PC52199-50MG
PC52199-100MG
APOSPC52199-50MGEA
0
EUR
APOSPC52199-50MG
APOSPC52199-100MG
5-(Difluoromethyl)pyridine-2-carbaldehyde 95%
5-(Difluoromethyl)pyridine-2-carbaldehyde
Formel:
C₇H₅F₂NO Molecular Weight: 157.118 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD18256104 CAS-Nummer: 955112-64-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...