4-Bromo-2-chloro-3-fluorobenzyl alcohol 95%

Lieferant: Apollo Scientific

PC52203-1G PC52203-250MG PC52203-5G
APOSPC52203-1GEA 416 EUR
APOSPC52203-1G APOSPC52203-250MG APOSPC52203-5G
4-Bromo-2-chloro-3-fluorobenzyl alcohol 95%
4-Bromo-2-chloro-3-fluorobenzyl alcohol
Formel: C₇H₅BrClFO
Molecular Weight: 239,469 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 1891698-71-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR