4-Bromo-2-chloro-3-fluoroacetophenone 95%
Lieferant: Apollo Scientific
PC52205-1G
PC52205-250MG
APOSPC52205-1GEA
560
EUR
APOSPC52205-1G
APOSPC52205-250MG
4-Bromo-2-chloro-3-fluoroacetophenone 95%
4-Bromo-2-chloro-3-fluoroacetophenone
Formel:
C₈H₅BrClFO Molecular Weight: 251,48 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD32695906 CAS-Nummer: 1898158-31-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...