4-Fluoro-2-(trifluoromethyl)-1H-indole 95%

Lieferant: Apollo Scientific

PC52225-1G PC52225-250MG PC52225-5G
APOSPC52225-1GEA 551 EUR
APOSPC52225-1G APOSPC52225-250MG APOSPC52225-5G
4-Fluoro-2-(trifluoromethyl)-1H-indole 95%
4-Fluoro-2-(trifluoromethyl)-1H-indole
Formel: C₉H₅F₄N
Molecular Weight: 203.136 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28526670
CAS-Nummer: 1780932-13-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR