3-Fluoro-2-iodo-N-methoxy-6-(trifluoromethyl)benzamide

Lieferant: Apollo Scientific

PC52449-1G PC52449-250MG
APOSPC52449-1GEA 574 EUR
APOSPC52449-1G APOSPC52449-250MG
3-Fluoro-2-iodo-N-methoxy-6-(trifluoromethyl)benzamide
3-Fluoro-2-iodo-N-methoxy-6-(trifluoromethyl)benzamide
Formel: C₉H₆F₄INO₂
Molecular Weight: 363.048 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2414146-33-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR